Assay Details
Target Gene Details
Entrez Gene ID: | 23640 |
Gene Name: | HSPA (Hsp70) binding protein 1 |
Gene Aliases: |
FES1 |
Location: |
Chr.19:55262222-55280383 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HSPBP1 | NM_001130106.1 | 9 | 1379 | NP_001123578.1 |
NM_001297600.1 | 7 | 1294 | NP_001284529.1 | |
NM_012267.4 | 8 | 1488 | NP_036399.3 | |
XM_005258700.2 | 8 | 1197 | XP_005258757.1 | |
XM_005258701.2 | 8 | 1259 | XP_005258758.1 | |
XM_011526691.1 | 9 | 1355 | XP_011524993.1 | |
XM_017026543.1 | 8 | 1194 | XP_016882032.1 | |
XM_017026544.1 | 8 | 1216 | XP_016882033.1 | |
XM_017026545.1 | 8 | 1208 | XP_016882034.1 | |
AB020592.1 | 8 | 1210 | BAB18742.1 | |
AF093420.1 | 8 | 1213 | AAC79703.1 | |
AF187859.1 | 8 | 1178 | AAF35833.1 | |
AF217996.1 | 9 | 1365 | AAG17238.1 | |
AI859141.1 | ||||
AK075293.1 | 8 | 1235 | ||
AK129755.1 | 8 | 1291 | ||
AK130781.1 | 8 | 1225 | ||
AK294358.1 | 7 | 1293 | ||
BC001236.1 | 7 | 1143 | AAH01236.1 | |
BC002373.2 | 8 | 1200 | AAH02373.1 | |
CR457118.1 | CAG33399.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |