Assay Details
Target Gene Details
Entrez Gene ID: | 9229 |
Gene Name: | DLG associated protein 1 |
Gene Aliases: |
DAP-1, DAP-1-ALPHA, DAP-1-BETA, DAP1, DLGAP1A, DLGAP1B, GKAP, SAPAP1 |
Location: |
Chr.18:3496032-4455307 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DLGAP1 | NM_001003809.2 | 10 | 2428 | NP_001003809.1 |
NM_001242762.1 | 10 | 2371 | NP_001229691.1 | |
NM_001242763.1 | 11 | 2485 | NP_001229692.1 | |
NM_001242764.1 | 10 | 2401 | NP_001229693.1 | |
NM_001242766.1 | 11 | 2458 | NP_001229695.1 | |
NM_001308390.1 | 11 | 2483 | NP_001295319.1 | |
NM_004746.3 | 13 | 3785 | NP_004737.2 | |
XM_005258171.2 | 14 | 3856 | XP_005258228.1 | |
XM_005258172.1 | 14 | 3851 | XP_005258229.1 | |
XM_005258173.3 | 12 | 2647 | XP_005258230.2 | |
XM_005258174.4 | 11 | 2704 | XP_005258231.1 | |
XM_006722367.3 | 12 | 2815 | XP_006722430.1 | |
XM_017026080.1 | 11 | 2784 | XP_016881569.1 | |
XM_017026081.1 | 10 | 2448 | XP_016881570.1 | |
XM_017026083.1 | 10 | 2673 | XP_016881572.1 | |
AB000276.1 | BAA23257.1 | |||
AB000277.1 | BAA23258.1 | |||
AK095761.1 | ||||
AK294717.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1059696 | Chr.18:3494893 - 3572306 on Build GRCh38 | Gain | DLGAP1 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |