Assay Details
Target Gene Details
Entrez Gene ID: | 2804 |
Gene Name: | golgin B1 |
Gene Aliases: |
GCP, GCP372, GOLIM1 |
Location: |
Chr.3:121663199-121749908 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 21 - Exon 21 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv8524n54 | Chr.3:121252053 - 121838686 on Build GRCh38 | Gain | EAF2 HCLS1 STXBP5L POLQ GOLGB1 IQCB1 FBXO40 ARGFX |
nsv829703 | Chr.3:121664443 - 121815646 on Build GRCh38 | Loss | GOLGB1 IQCB1 |
esv3575599 | Chr.3:121571453 - 121671424 on Build GRCh38 | Gain | HCLS1 GOLGB1 FBXO40 ARGFX |
nsv998740 | Chr.3:121570652 - 121671424 on Build GRCh38 | Gain | HCLS1 GOLGB1 FBXO40 ARGFX |
esv3575598 | Chr.3:121406926 - 121715587 on Build GRCh38 | Gain | HCLS1 STXBP5L POLQ GOLGB1 FBXO40 ARGFX |
nsv999335 | Chr.3:121381436 - 121689919 on Build GRCh38 | Gain | HCLS1 STXBP5L POLQ GOLGB1 FBXO40 ARGFX |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |