Assay Details
Target Gene Details
Entrez Gene ID: | 285033 |
Gene Name: | STARD7 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.2:96208416-96242623 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
STARD7-AS1 | NR_046322.1 | 3 | 1328 | |
AK057419.1 | 3 | 800 | ||
AK311432.1 | 3 | 1343 | ||
BC101730.1 | 1 | 43 | AAI01731.1 | |
BC101732.1 | 1 | 43 | AAI01733.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834305 | Chr.2:96183160 - 96353738 on Build GRCh38 | Gain | STARD7-AS1 TMEM127 SNRNP200 ITPRIPL1 CIAO1 NCAPH STARD7 LOC105373495 |
dgv607e201 | Chr.2:95855884 - 97561622 on Build GRCh38 | Deletion | CNNM4 NEURL3 STARD7 FAHD2B KANSL3 MIR3127 STARD7-AS1 ADRA2B CIAO1 NCAPH CNNM3 ASTL LOC105373496 ANKRD36B DUSP2 ANKRD39 LOC100506123 LOC101927053 LOC100506076 FER1L5 SEMA4C LOC653924 FAHD2CP ANKRD36C TMEM127 SNRNP200 ITPRIPL1 ARID5A LOC107984110 FAM178B ANKRD23 ANKRD36 LOC105373495 LMAN2L GPAT2 |
nsv1135873 | Chr.2:95981454 - 97579645 on Build GRCh38 | Deletion | CNNM4 NEURL3 STARD7 FAHD2B KANSL3 MIR3127 STARD7-AS1 ADRA2B CIAO1 NCAPH CNNM3 ASTL LOC105373496 ANKRD36B DUSP2 ANKRD39 LOC100506123 LOC101927053 LOC100506076 FER1L5 SEMA4C LOC653924 FAHD2CP ANKRD36C TMEM127 SNRNP200 ITPRIPL1 ARID5A LOC107984110 FAM178B ANKRD23 ANKRD36 LOC105373495 LMAN2L GPAT2 |
esv3425717 | Chr.2:95859900 - 97515905 on Build GRCh38 | Duplication | CNNM4 NEURL3 STARD7 FAHD2B KANSL3 MIR3127 STARD7-AS1 ADRA2B CIAO1 NCAPH CNNM3 ASTL LOC105373496 ANKRD36B DUSP2 ANKRD39 LOC100506123 LOC101927053 LOC100506076 FER1L5 SEMA4C LOC653924 FAHD2CP ANKRD36C TMEM127 SNRNP200 ITPRIPL1 ARID5A LOC107984110 FAM178B ANKRD23 ANKRD36 LOC105373495 LMAN2L GPAT2 |