Assay Details
Target Gene Details
Entrez Gene ID: | 4793 |
Gene Name: | NFKB inhibitor beta |
Gene Aliases: |
IKBB, TRIP9 |
Location: |
Chr.19:38899503-38908894 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NFKBIB | NM_001243116.1 | 4 | 536 | NP_001230045.1 |
NM_002503.4 | 4 | 778 | NP_002494.2 | |
NR_040515.1 | 4 | 722 | ||
XM_006723226.2 | 4 | 714 | XP_006723289.2 | |
XM_006723227.3 | 4 | 677 | XP_006723290.3 | |
AB209784.1 | 4 | 744 | BAD93021.1 | |
AI684894.1 | 3 | 466 | ||
AK057862.1 | 4 | 673 | BAB71599.1 | |
AK290569.1 | 4 | 729 | ||
BC007197.1 | 2 | 378 | AAH07197.1 | |
BC015528.1 | 4 | 709 | AAH15528.1 | |
BI520169.1 | 4 | 536 | ||
BM449613.1 | 4 | 733 | ||
BQ440907.1 | 4 | 510 | ||
BT006743.1 | 4 | 675 | AAP35389.1 | |
BU150265.1 | ||||
BU634404.1 | 3 | 474 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |