Assay Details
Target Gene Details
Entrez Gene ID: | 58506 |
Gene Name: | SR-related CTD associated factor 1 |
Gene Aliases: |
SRA1 |
Location: |
Chr.19:49640483-49658649 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SCAF1 | NM_021228.2 | NP_067051.2 | ||
XM_005259122.4 | XP_005259179.1 | |||
XM_011527194.2 | XP_011525496.1 | |||
AK024444.1 | ||||
BC011662.2 | AAH11662.2 | |||
BC053992.1 | AAH53992.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv469715 | Chr.19:49572544 - 49730286 on Build GRCh38 | Gain+Loss | NOSIP BCL2L12 SCAF1 CPT1C PRMT1 RRAS PRRG2 PRR12 MIR5088 ADM5 IRF3 |
nsv833865 | Chr.19:49572545 - 49730286 on Build GRCh38 | Gain+Loss | NOSIP BCL2L12 SCAF1 CPT1C PRMT1 RRAS PRRG2 PRR12 MIR5088 ADM5 IRF3 |
nsv9739 | Chr.19:47901840 - 50143063 on Build GRCh38 | Gain+Loss | SPACA4 MAMSTR IZUMO1 TBC1D17 SNORD35A GRIN2D KCNA7 PLA2G4C-AS1 ZNF114 FCGRT SNAR-C1 SNAR-A12 TSKS CA11 SEC1P SNAR-A8 LIG1 SIGLEC11 PLEKHA4 SLC17A7 SNAR-A5 SNAR-C5 VRK3 RPL18 LOC100287477 RASIP1 NOSIP SNAR-A14 SNAR-A3 MED25 SNAR-A7 LMTK3 GRWD1 FUZ CYTH2 MIR4750 LOC101059948 NUP62 MIR4324 MIR4749 SNAR-A1 SLC6A16 PRRG2 MIR6799 LOC105447645 FAM83E SULT2B1 TMEM143 PRR12 SNAR-G2 SNAR-B1 BSPH1 RPL13A BCL2L12 NUCB1 KCNJ14 SNORD33 TEAD2 MIR4751 FUT2 SNORD32A SNAR-A11 MIR5088 IRF3 LOC105372430 MIR6798 NTN5 PLA2G4C SNAR-B2 PTOV1-AS2 CGB5 SNAR-C4 CARD8-AS1 SNAR-C2 CABP5 C19orf68 LOC101928295 SCAF1 GYS1 HSD17B14 RCN3 NTF4 AKT1S1 BCAT2 FLT3LG ZNF473 SIGLEC16 MIR150 SNAR-A2 LIN7B SNAR-G1 CCDC155 FGF21 TRPM4 SNAR-C3 LHB ALDH16A1 PTH2 PTOV1-AS1 ELSPBP1 RRAS PNKP PPP1R15A FUT1 DHDH MIR6800 FTL ATF5 SYNGR4 SNAR-A6 SNORD34 HRC SNAR-A13 CPT1C PRMT1 SNAR-D PPFIA3 NUCB1-AS1 SNRNP70 EMP3 AP2A1 C19orf73 SNAR-A4 CGB7 CGB2 SPHK2 DKKL1 CD37 CARD8 SNORD35B IL4I1 SNAR-A10 CGB8 PTOV1 PIH1D1 DBP SNAR-A9 CCDC114 RPS11 CGB1 FLJ26850 RUVBL2 ADM5 GFY CGB3 KDELR1 BAX TULP2 |
nsv817854 | Chr.19:49618821 - 49726387 on Build GRCh38 | Gain | BCL2L12 SCAF1 CPT1C PRMT1 RRAS PRR12 MIR5088 ADM5 IRF3 |
nsv953600 | Chr.19:49555044 - 49938043 on Build GRCh38 | Deletion | MIR4749 SCAF1 CPT1C TBC1D17 PRMT1 PRRG2 AKT1S1 MIR6799 AP2A1 TSKS PRR12 IL4I1 PTOV1 BCL2L12 MIR4751 PTOV1-AS1 RRAS PNKP MIR5088 ADM5 IRF3 NOSIP MIR6800 MED25 ATF5 PTOV1-AS2 FUZ MIR4750 NUP62 |
nsv833864 | Chr.19:49551790 - 49733819 on Build GRCh38 | Gain | NOSIP BCL2L12 SCAF1 CPT1C PRMT1 RRAS PRRG2 PRR12 MIR5088 ADM5 IRF3 |
nsv1160655 | Chr.19:49489110 - 49841830 on Build GRCh38 | Deletion | SNORD34 SCAF1 CPT1C PRMT1 RCN3 SNORD35A PRRG2 MIR6799 MIR150 FCGRT AP2A1 TSKS PRR12 SNORD35B RPL13A BCL2L12 SNORD33 PTOV1-AS1 SNORD32A RRAS RPS11 MIR5088 ADM5 IRF3 NOSIP MIR6800 MED25 FUZ |
nsv828578 | Chr.19:49560777 - 49680548 on Build GRCh38 | Loss | NOSIP BCL2L12 SCAF1 PRMT1 RRAS PRRG2 PRR12 IRF3 |
nsv579908 | Chr.19:49645135 - 49663393 on Build GRCh38 | Loss | SCAF1 IRF3 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |