Assay Details
Target Gene Details
Entrez Gene ID: | 23640 |
Gene Name: | HSPA (Hsp70) binding protein 1 |
Gene Aliases: |
FES1 |
Location: |
Chr.19:55262222-55280383 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HSPBP1 | NM_001130106.1 | 4 | 631 | NP_001123578.1 |
NM_001297600.1 | 2 | 546 | NP_001284529.1 | |
NM_012267.4 | 3 | 740 | NP_036399.3 | |
XM_005258700.2 | 3 | 471 | XP_005258757.1 | |
XM_005258701.2 | 3 | 533 | XP_005258758.1 | |
XM_011526691.1 | 4 | 629 | XP_011524993.1 | |
XM_017026543.1 | 3 | 468 | XP_016882032.1 | |
XM_017026544.1 | 3 | 468 | XP_016882033.1 | |
XM_017026545.1 | 3 | 471 | XP_016882034.1 | |
AB020592.1 | 3 | 462 | BAB18742.1 | |
AF093420.1 | 3 | 465 | AAC79703.1 | |
AF187859.1 | 3 | 430 | AAF35833.1 | |
AF217996.1 | 4 | 617 | AAG17238.1 | |
AK075293.1 | 3 | 487 | ||
AK129755.1 | 3 | 543 | ||
AK130781.1 | 3 | 477 | ||
AK294358.1 | 2 | 545 | ||
BC001236.1 | 2 | 395 | AAH01236.1 | |
BC002373.2 | 3 | 452 | AAH02373.1 | |
CR457118.1 | 2 | 297 | CAG33399.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |