Assay Details
Target Gene Details
Entrez Gene ID: | 57568 |
Gene Name: | signal induced proliferation associated 1 like 2 |
Gene Aliases: |
SPAL2 |
Location: |
Chr.1:232397965-232630491 on Build GRCh38 |
Assay Gene Location: | Within Exon 25 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SIPA1L2 | NM_020808.4 | 21 | 5679 | NP_065859.3 |
XM_005273211.2 | 23 | 6016 | XP_005273268.1 | |
XM_005273212.4 | 23 | 5823 | XP_005273269.1 | |
XM_005273213.4 | 22 | 5716 | XP_005273270.1 | |
XM_011544243.2 | 24 | 5872 | XP_011542545.1 | |
XM_017001896.1 | 23 | 5769 | XP_016857385.1 | |
XM_017001897.1 | 22 | 5962 | XP_016857386.1 | |
XM_017001898.1 | 22 | 5736 | XP_016857387.1 | |
AB037810.1 | 21 | 4883 | BAA92627.1 | |
AK022852.1 | 12 | 2225 | BAB14273.1 | |
AK026779.1 | 8 | 1651 | ||
AK074212.1 | 6 | 1112 | ||
AK093191.1 | 14 | 2761 | BAC04090.1 | |
AY168879.1 | 21 | 5639 | AAO12530.1 | |
BC013119.1 | 12 | 2136 | ||
BC082251.1 | 12 | 2165 | AAH82251.1 | |
BC150334.1 | 21 | 4894 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1006763 | Chr.1:232341238 - 232458244 on Build GRCh38 | Gain | SIPA1L2 |
nsv549355 | Chr.1:232388426 - 232435636 on Build GRCh38 | Gain | SIPA1L2 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |