Assay Details
Target Gene Details
Entrez Gene ID: | 54972 |
Gene Name: | transmembrane protein 132A |
Gene Aliases: |
GBP, HSPA5BP1 |
Location: |
Chr.11:60924441-60937159 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TMEM132A | NM_017870.3 | 4 | 1004 | NP_060340.2 |
NM_178031.2 | 4 | 1004 | NP_821174.1 | |
XM_017017951.1 | 3 | 2601 | XP_016873440.1 | |
XM_017017952.1 | 3 | 2600 | XP_016873441.1 | |
AB046803.1 | 3 | 847 | BAB13409.1 | |
AK000546.1 | BAA91245.1 | |||
AL359557.2 | 1 | 252 | CAH10668.2 | |
AL834133.1 | 3 | 866 | ||
BC028106.1 | 4 | 1203 | AAH28106.1 | |
BC032059.1 | 4 | 964 | AAH32059.1 | |
BC114569.1 | 4 | 969 | AAI14570.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |