Assay Details
Target Gene Details
Entrez Gene ID: | 10845 |
Gene Name: | caseinolytic mitochondrial matrix peptidase chaperone subunit |
Gene Aliases: |
- |
Location: |
Chr.15:65148219-65185420 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CLPX | NM_006660.4 | 5 | 900 | NP_006651.2 |
NR_133680.1 | 5 | 900 | ||
XM_011521164.2 | 5 | 884 | XP_011519466.1 | |
AJ006267.3 | 5 | 705 | CAA06933.2 | |
AK292316.1 | 5 | 826 | ||
AK308133.1 | 5 | 820 | ||
AK310614.1 | 5 | 822 | ||
AL136922.1 | 5 | 803 | CAB66856.1 | |
BC130373.1 | 5 | 696 | ||
BC136487.1 | 5 | 696 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv569704 | Chr.15:65162973 - 65178902 on Build GRCh38 | Loss | CLPX |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |