Assay Details
Target Gene Details
Entrez Gene ID: | 57585 |
Gene Name: | cramped chromatin regulator homolog 1 |
Gene Aliases: |
CRAMP1L, HN1L, TCE4 |
Location: |
Chr.16:1614640-1677908 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CRAMP1 | NM_020825.3 | NP_065876.3 | ||
AB037847.2 | BAA92664.2 | |||
BC137172.1 | ||||
BC137173.1 | ||||
BC152477.1 | ||||
BE048705.1 | ||||
BM451163.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv4856n54 | Chr.16:1600945 - 1813519 on Build GRCh38 | Loss | EME2 NME3 NUBP2 MIR3177 IFT140 CRAMP1 SPSB3 IGFALS HAGH MRPS34 MAPK8IP3 LOC105371046 HN1L |
esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication | POLR3K PIGQ MIR3176 HBA1 HBA2 MIR3177 CACNA1H MIR662 RNPS1 ATP6V0C TMEM204 METRN JMJD8 PRSS27 IGFALS SNORD60 RHBDL1 NPRL3 E4F1 TBC1D24 TBL3 SNHG9 LOC652276 LOC105371038 RPUSD1 RAB11FIP3 GNG13 BRICD5 LOC100287175 LOC105371184 MIR3178 NUBP2 LOC107987233 MSLN CRAMP1 PKD1 TPSG1 HAGH RHOT2 LOC729652 HN1L PGP PTX4 LOC100134368 DNASE1L2 TELO2 SLC9A3R2 LINC00254 TPSB2 C16orf91 MEIOB CCDC78 MIR940 FBXL16 ERVK13-1 MIR1225 NARFL IFT140 LMF1 UNKL MSRB1 WFIKKN1 MRPS34 TMEM8A LUC7L WDR24 SYNGR3 TSR3 SNRNP25 CCNF DECR2 RAB26 TRAF7 MIR6511B1 CAPN15 HBQ1 PDPK1 SNORA78 ARHGDIG MIR3677 BAIAP3 PDIA2 LOC107984876 MIR6768 CEMP1 RPL23AP5 RHBDF1 NDUFB10 SNORA64 MIR4516 SOX8 PRR35 NTN3 RAB40C WASIR2 HBZ NHLRC4 AXIN1 CASKIN1 UBE2I HBM HAGHL NPW MAPK8IP3 RPL3L MPG GFER MCRIP2 MIR6859-4 MLST8 EME2 MIR3180-5 LMF1-AS1 FAM234A LINC00235 MIR4717 TSC2 ABCA3 MIR6767 LOC105371046 ZNF598 C16orf59 LOC101929280 LOC106660606 NME4 MRPL28 AMDHD2 CHTF18 SNHG19 FAHD1 LOC105371045 NME3 C1QTNF8 TPSD1 NOXO1 METTL26 WDR90 RNF151 NTHL1 ABCA17P KCTD5 PRR25 PRSS29P RGS11 STUB1 SSTR5 SNORA10 TPSAB1 ECI1 HS3ST6 RPS2 FLJ42627 LOC105371049 SSTR5-AS1 FAM173A DDX11L10 SPSB3 LOC105371047 MIR5587 CLCN7 GNPTG CCDC154 |
nsv1051692 | Chr.16:1505070 - 1866102 on Build GRCh38 | Loss | TELO2 MEIOB EME2 FAHD1 NME3 NUBP2 MIR3177 IFT140 TMEM204 CRAMP1 SPSB3 LOC105371047 IGFALS HAGH MRPS34 MAPK8IP3 LOC105371046 HN1L |
dgv4855n54 | Chr.16:1577507 - 1694025 on Build GRCh38 | Loss | CRAMP1 LOC105371046 HN1L IFT140 |
nsv428317 | Chr.16:1550716 - 1711581 on Build GRCh38 | Gain | TMEM204 CRAMP1 MAPK8IP3 LOC105371046 HN1L IFT140 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |