Assay Details
Target Gene Details
Entrez Gene ID: | 51005 |
Gene Name: | amidohydrolase domain containing 2 |
Gene Aliases: |
CGI-14 |
Location: |
Chr.16:2520351-2531414 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AMDHD2 | NM_001145815.1 | NP_001139287.1 | ||
NM_015944.3 | NP_057028.2 | |||
XM_017023262.1 | XP_016878751.1 | |||
XM_017023263.1 | XP_016878752.1 | |||
XM_017023264.1 | XP_016878753.1 | |||
XM_017023265.1 | XP_016878754.1 | |||
XM_017023266.1 | XP_016878755.1 | |||
XM_017023267.1 | XP_016878756.1 | |||
AB209351.1 | 2 | 3582 | BAD92588.1 | |
AF132948.1 | AAD27723.1 | |||
AK296877.1 | ||||
AK300747.1 | ||||
BC018734.1 | AAH18734.1 | |||
CN362870.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication | MSRB1 LOC100287175 MIR940 SNORA64 WFIKKN1 MIR4717 DDX11L10 PTX4 SYNGR3 ZNF598 RNF151 CHTF18 RAB11FIP3 MEIOB RAB40C CAPN15 POLR3K SOX8 NARFL SSTR5-AS1 CEMP1 TMEM8A MIR6859-4 PRSS27 C16orf59 HBM CLCN7 RNPS1 LUC7L TPSD1 TPSB2 MIR6511B1 NME4 FAM173A LOC107987233 DECR2 PGP CCDC154 TELO2 KCTD5 NTN3 LOC101929280 HAGH DNASE1L2 PKD1 TBL3 LOC105371184 CASKIN1 MIR4516 HBZ SNORD60 LOC105371045 ERVK13-1 METRN PIGQ RHOT2 TRAF7 LOC105371047 ATP6V0C MIR3176 RPS2 SSTR5 WDR90 MPG ARHGDIG LOC106660606 NDUFB10 MIR6767 AMDHD2 TSR3 MIR5587 E4F1 RHBDF1 LOC100134368 STUB1 LOC105371049 MSLN LMF1-AS1 FAM234A AXIN1 RPUSD1 MIR3677 SNORA10 NHLRC4 BRICD5 EME2 SNRNP25 UBE2I LOC105371038 GNPTG TMEM204 CCNF MIR1225 PRR35 LMF1 WDR24 TBC1D24 C1QTNF8 HS3ST6 MAPK8IP3 HAGHL LINC00235 UNKL RPL23AP5 NUBP2 PRR25 CRAMP1 PRSS29P RPL3L FBXL16 IGFALS SNORA78 ABCA17P PDPK1 TSC2 LOC729652 PDIA2 MRPL28 FAHD1 LOC652276 NTHL1 SNHG9 GNG13 RGS11 LINC00254 MCRIP2 MIR6768 NPW SLC9A3R2 NPRL3 SPSB3 FLJ42627 LOC107984876 ABCA3 MIR3180-5 LOC105371046 MIR3177 MLST8 HBA2 ECI1 C16orf91 RAB26 MIR662 JMJD8 CACNA1H IFT140 TPSAB1 CCDC78 HBQ1 HN1L NOXO1 NME3 TPSG1 BAIAP3 HBA1 GFER RHBDL1 METTL26 MRPS34 WASIR2 SNHG19 MIR3178 |
esv2760067 | Chr.16:2486824 - 2820535 on Build GRCh38 | Gain+Loss | ERVK13-1 CEMP1 KCTD5 PRSS21 SRRM2-AS1 PRSS27 ATP6V0C FLJ42627 PRSS33 TCEB2 PRSS41 SRRM2 PDPK1 AMDHD2 TBC1D24 LOC652276 MIR3178 |
nsv833122 | Chr.16:2501717 - 2663937 on Build GRCh38 | Loss | ERVK13-1 CEMP1 PDPK1 AMDHD2 TBC1D24 ATP6V0C LOC652276 FLJ42627 MIR3178 |
nsv1052260 | Chr.16:2074546 - 2629655 on Build GRCh38 | Gain | PGP MIR3677 MIR940 BRICD5 NTN3 MIR6768 DNASE1L2 MIR4717 PKD1 ABCA3 CCNF MIR3180-5 MIR1225 MLST8 CASKIN1 TBC1D24 MIR4516 ECI1 RAB26 SNORD60 CEMP1 TRAF7 C16orf59 ATP6V0C RNPS1 LOC106660606 MIR6511B1 ABCA17P PDPK1 MIR6767 AMDHD2 TSC2 LOC729652 E4F1 LOC652276 LOC105371049 SNHG19 MIR3178 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |