Assay Details
Target Gene Details
Entrez Gene ID: | 2026 |
Gene Name: | enolase 2 |
Gene Aliases: |
HEL-S-279, NSE |
Location: |
Chr.12:6914450-6923696 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 11 - Intron 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ENO2 | NM_001975.2 | NP_001966.1 | ||
AK124656.1 | ||||
AK290525.1 | ||||
AK295220.1 | ||||
AK312402.1 | ||||
BC002745.2 | AAH02745.1 | |||
BT007383.1 | AAP36047.1 | |||
CR536582.1 | CAG38819.1 | |||
EU794607.1 | ||||
M22349.1 | AAB59554.1 | |||
M36768.1 | AAA52388.1 | |||
X13120.1 | CAA31512.1 | |||
X14327.1 | CAA32505.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1047373 | Chr.12:6902891 - 7055754 on Build GRCh38 | Gain | LOC105369635 PTPN6 RNU7-1 EMG1 C12orf57 ATN1 SCARNA12 LRRC23 PHB2 MIR200C MIR141 LPCAT3 ENO2 |
nsv509453 | Chr.12:6896530 - 7007852 on Build GRCh38 | Insertion | LOC105369635 PTPN6 RNU7-1 EMG1 C12orf57 ATN1 SCARNA12 LRRC23 PHB2 MIR200C MIR141 LPCAT3 ENO2 |
nsv557261 | Chr.12:6839304 - 6924660 on Build GRCh38 | Loss | ATN1 RPL13P5 LOC105369632 DSTNP2 GNB3 LRRC23 USP5 CDCA3 ENO2 P3H3 SPSB2 TPI1 |
nsv832324 | Chr.12:6780297 - 6932306 on Build GRCh38 | Loss | RPL13P5 P3H3 SPSB2 TPI1 GPR162 CD4 ATN1 LOC105369632 DSTNP2 GNB3 LRRC23 USP5 CDCA3 ENO2 |
nsv1035811 | Chr.12:6876316 - 6994911 on Build GRCh38 | Gain | LOC105369635 RPL13P5 PTPN6 RNU7-1 EMG1 C12orf57 ATN1 SCARNA12 LOC105369632 DSTNP2 LRRC23 PHB2 MIR200C MIR141 LPCAT3 ENO2 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |