Assay Details
Target Gene Details
Entrez Gene ID: | 2217 |
Gene Name: | Fc fragment of IgG receptor and transporter |
Gene Aliases: |
FCRN, alpha-chain |
Location: |
Chr.19:49512279-49526428 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FCGRT | NM_001136019.2 | 7 | 1575 | NP_001129491.1 |
NM_004107.4 | 6 | 1311 | NP_004098.1 | |
XM_005258657.1 | 6 | 1130 | XP_005258714.1 | |
XM_011526627.2 | 5 | 992 | XP_011524929.1 | |
AK074734.1 | 3 | 2453 | ||
AK075510.1 | 7 | 1509 | ||
AK075532.1 | 7 | 1225 | ||
BC008734.2 | 6 | 1255 | AAH08734.1 | |
BE245284.1 | 3 | 345 | ||
BT006828.1 | AAP35474.1 | |||
BX647163.1 | 7 | 1195 | CAI46032.1 | |
U12255.1 | 6 | 1214 | AAA58958.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv9739 | Chr.19:47901840 - 50143063 on Build GRCh38 | Gain+Loss | FLJ26850 MIR4749 SNAR-A8 CPT1C TBC1D17 MIR4750 SNAR-A7 NUP62 SNORD35B FGF21 PPFIA3 MIR6799 BCL2L12 SNAR-A9 SNAR-G2 SPHK2 CGB2 SNAR-A10 PTOV1-AS1 SNAR-C1 NTN5 CGB8 IZUMO1 TSKS KCNJ14 FUZ ELSPBP1 ALDH16A1 VRK3 SIGLEC11 FUT1 SNAR-A5 RPL13A NUCB1 SNAR-A14 SNAR-C2 MIR150 LMTK3 NOSIP EMP3 SYNGR4 RPS11 HSD17B14 TULP2 PRR12 PLA2G4C-AS1 SNAR-C3 CABP5 CCDC155 IRF3 SPACA4 CARD8 SNAR-A13 SNAR-A2 MIR5088 ADM5 RCN3 LIN7B MAMSTR PTOV1 SNAR-A11 SNORD32A RRAS DBP SNORD34 PNKP AP2A1 SNAR-A1 PIH1D1 FUT2 CCDC114 MIR6800 LHB ZNF473 LOC100287477 MIR4751 CARD8-AS1 CGB1 SNAR-A3 SIGLEC16 SULT2B1 SNAR-A6 SNRNP70 KDELR1 PRRG2 MIR6798 RPL18 DKKL1 SLC17A7 CGB3 GYS1 FCGRT SNAR-G1 KCNA7 BSPH1 PTH2 ZNF114 PLA2G4C CYTH2 LOC105372430 TMEM143 TRPM4 RUVBL2 SNAR-C5 FLT3LG C19orf68 NUCB1-AS1 TEAD2 PRMT1 MED25 GRIN2D SNAR-A4 CD37 AKT1S1 LIG1 SNAR-C4 SNAR-B1 RASIP1 FTL SNAR-D CA11 SCAF1 SNORD35A SNAR-B2 MIR4324 DHDH GFY BAX SNORD33 PLEKHA4 BCAT2 C19orf73 IL4I1 SLC6A16 LOC105447645 CGB7 ATF5 CGB5 NTF4 LOC101928295 LOC101059948 FAM83E SNAR-A12 GRWD1 SEC1P PPP1R15A HRC PTOV1-AS2 |
nsv833863 | Chr.19:49479177 - 49629716 on Build GRCh38 | Loss | SNORD32A PRRG2 PRR12 SNORD34 SNORD35A FLT3LG SNORD35B SNORD33 RPL13A FCGRT MIR150 NOSIP RCN3 RPS11 |
nsv833862 | Chr.19:49448789 - 49570676 on Build GRCh38 | Loss | SNORD32A SNORD34 SNORD35A PIH1D1 FLT3LG SNORD35B SNORD33 ALDH16A1 RPL13A FCGRT MIR150 NOSIP RCN3 RPS11 |
nsv1160655 | Chr.19:49489110 - 49841830 on Build GRCh38 | Deletion | PRRG2 CPT1C PRR12 SCAF1 SNORD35A SNORD35B IRF3 SNORD33 MIR6799 FCGRT MIR5088 ADM5 BCL2L12 RCN3 SNORD32A PTOV1-AS1 RRAS SNORD34 AP2A1 TSKS FUZ MIR6800 RPL13A MIR150 NOSIP PRMT1 MED25 RPS11 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |