Assay Details
Target Gene Details
Entrez Gene ID: | 10146 |
Gene Name: | G3BP stress granule assembly factor 1 |
Gene Aliases: |
G3BP, HDH-VIII |
Location: |
Chr.5:151771915-151811352 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
G3BP1 | NM_005754.2 | 7 | 753 | NP_005745.1 |
NM_198395.1 | 7 | 722 | NP_938405.1 | |
XM_006714749.3 | 7 | 742 | XP_006714812.1 | |
XM_006714750.3 | 7 | 681 | XP_006714813.1 | |
XM_017008922.1 | 7 | 1028 | XP_016864411.1 | |
XM_017008923.1 | 7 | 997 | XP_016864412.1 | |
AK130003.1 | 5 | 438 | BAC85272.1 | |
AK222606.1 | 7 | 714 | BAD96326.1 | |
AK300098.1 | 6 | 623 | ||
AK303580.1 | 7 | 810 | ||
AK312572.1 | 7 | 716 | ||
BC006997.1 | 7 | 716 | AAH06997.1 | |
BC108278.1 | 7 | 661 | AAI08279.1 | |
BT019384.1 | 6 | 582 | AAV38191.1 | |
BU517281.1 | 7 | 746 | ||
BX647869.1 | 6 | 624 | CAI46065.1 | |
BX648012.1 | 7 | 728 | CAI46067.1 | |
CR536535.1 | 6 | 582 | CAG38772.1 | |
U32519.1 | 7 | 703 | AAB07787.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |