Assay Details
Target Gene Details
Entrez Gene ID: | 64689 |
Gene Name: | golgi reassembly stacking protein 1 |
Gene Aliases: |
GOLPH5, GRASP65, P65 |
Location: |
Chr.3:39096599-39108363 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GORASP1 | NM_001278789.1 | 7 | 1267 | NP_001265718.1 |
NM_001278790.1 | 6 | 1087 | NP_001265719.1 | |
NM_031899.3 | 9 | 2015 | NP_114105.1 | |
NR_103866.1 | 9 | 1511 | ||
NR_103867.1 | 9 | 1503 | ||
XM_006713301.3 | 9 | 2115 | XP_006713364.1 | |
XM_011534020.2 | 9 | 2025 | XP_011532322.1 | |
XM_011534021.2 | 8 | 1935 | XP_011532323.1 | |
XM_017007050.1 | 9 | 1201 | XP_016862539.1 | |
XM_017007051.1 | 8 | 1550 | XP_016862540.1 | |
XM_017007052.1 | 8 | 1146 | XP_016862541.1 | |
AJ409349.1 | 9 | 1291 | CAC35160.1 | |
AK001574.1 | 8 | 1533 | ||
AK057043.1 | 6 | 825 | ||
AK091168.1 | 9 | 1439 | BAC03598.1 | |
AK124755.1 | 9 | 2015 | ||
AK303847.1 | 7 | 1007 | ||
BC008928.2 | 9 | 1232 | AAH08928.1 | |
BC075854.1 | 9 | 1301 | AAH75854.1 | |
BC103826.1 | 9 | 1233 | AAI03827.1 | |
BC103827.1 | 9 | 1233 | AAI03828.1 | |
BM470731.1 | 4 | 528 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |