Assay Details
Target Gene Details
Entrez Gene ID: | 1355 |
Gene Name: | COX15, cytochrome c oxidase assembly homolog |
Gene Aliases: |
CEMCOX2 |
Location: |
Chr.10:99694793-99732667 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
COX15 | NM_001320974.1 | 1 | 336 | NP_001307903.1 |
NM_001320975.1 | 1 | 336 | NP_001307904.1 | |
NM_001320976.1 | 1 | 336 | NP_001307905.1 | |
NM_004376.6 | 1 | 336 | NP_004367.2 | |
NM_078470.5 | 1 | 336 | NP_510870.1 | |
XM_017015715.1 | 1 | 337 | XP_016871204.1 | |
BX537557.1 | 1 | 336 | CAD97781.1 | |
DB094258.1 | 1 | 337 |
Target Gene Details
Entrez Gene ID: | 51076 |
Gene Name: | cutC copper transporter |
Gene Aliases: |
CGI-32 |
Location: |
Chr.10:99732201-99756137 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CUTC | NM_015960.2 | 1 | 131 | NP_057044.2 |
AF132966.1 | 1 | 86 | AAD27741.1 | |
AK001142.1 | 1 | 51 | ||
AK302507.1 | 1 | 67 | ||
AK314687.1 | 1 | 67 | ||
BC021105.1 | AAH21105.1 | |||
BC028948.1 | 1 | 99 | AAH28948.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |