Assay Details
Target Gene Details
Entrez Gene ID: | 51247 |
Gene Name: | poly(A) binding protein interacting protein 2 |
Gene Aliases: |
PAIP-2, PAIP2A |
Location: |
Chr.5:139341832-139369720 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PAIP2 | NM_001033112.2 | NP_001028284.1 | ||
NM_016480.4 | NP_057564.3 | |||
NR_109830.1 | ||||
XM_011543428.1 | XP_011541730.1 | |||
XM_017009522.1 | XP_016865011.1 | |||
XM_017009523.1 | XP_016865012.1 | |||
AF151052.1 | AAF36138.1 | |||
AF317675.1 | AAK11562.1 | |||
AK024552.1 | ||||
AK314672.1 | ||||
AL136640.1 | CAB66575.1 | |||
BC001716.1 | AAH01716.1 | |||
BC039337.1 | AAH39337.1 | |||
BC048106.1 | AAH48106.1 | |||
BC062718.1 | AAH62718.1 | |||
BI552800.1 | ||||
BI560379.1 | ||||
CR533489.1 | CAG38520.1 |
Target Gene Details
Entrez Gene ID: | 9963 |
Gene Name: | solute carrier family 23 member 1 |
Gene Aliases: |
SLC23A2, SVCT1, YSPL3 |
Location: |
Chr.5:139367196-139384553 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC23A1 | NM_005847.4 | NP_005838.3 | ||
NM_152685.3 | NP_689898.2 | |||
XM_005272148.3 | XP_005272205.3 | |||
XM_005272149.3 | XP_005272206.3 | |||
XM_006714741.2 | XP_006714804.2 | |||
XM_011543766.1 | XP_011542068.1 | |||
XM_011543767.1 | XP_011542069.1 | |||
XM_011543768.1 | XP_011542070.1 | |||
XM_011543769.1 | XP_011542071.1 | |||
AF098277.1 | AAF22490.1 | |||
AF170911.1 | AAF24759.1 | |||
AK092655.1 | ||||
AK290789.1 | ||||
BC019225.1 | AAH19225.1 | |||
BC050261.1 | AAH50261.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |