Assay Details
Target Gene Details
Entrez Gene ID: | 150223 |
Gene Name: | YdjC homolog (bacterial) |
Gene Aliases: |
- |
Location: |
Chr.22:21628089-21630326 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
YDJC | NM_001017964.1 | 5 | 1059 | NP_001017964.1 |
XM_011529906.2 | 4 | 1156 | XP_011528208.1 | |
BC110075.1 | 4 | 865 | AAI10076.1 | |
BC133043.1 | 5 | 1213 | ||
BC133047.1 | 5 | 1213 | ||
BQ050386.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1160730 | Chr.22:21373849 - 21633730 on Build GRCh38 | Deletion | CCDC116 RIMBP3C YDJC PI4KAP2 HIC2 UBE2L3 RIMBP3B TMEM191C |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |