Assay Details
Target Gene Details
Entrez Gene ID: | 10397 |
Gene Name: | N-myc downstream regulated 1 |
Gene Aliases: |
CAP43, CMT4D, DRG-1, DRG1, GC4, HMSNL, NDR1, NMSL, PROXY1, RIT42, RTP, TARG1, TDD5 |
Location: |
Chr.8:133237171-133297587 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NDRG1 | NM_001135242.1 | 16 | 3039 | NP_001128714.1 |
NM_001258432.1 | 14 | 2525 | NP_001245361.1 | |
NM_001258433.1 | 15 | 2536 | NP_001245362.1 | |
NM_006096.3 | 16 | 2642 | NP_006087.2 | |
XM_011516791.1 | 16 | 2976 | XP_011515093.1 | |
XM_011516792.1 | 9 | 2875 | XP_011515094.1 | |
AF004162.1 | 15 | 2481 | AAC13419.1 | |
AK091147.1 | 15 | 2485 | ||
AK091269.1 | ||||
AK095649.1 | 14 | 2378 | BAC04597.1 | |
AK096562.1 | 16 | 2477 | ||
AK124709.1 | 8 | 3549 | ||
AK126924.1 | 16 | 3321 | ||
AK129867.1 | 7 | 1916 | ||
AK130985.1 | 1 | 165 | ||
BC003175.1 | 16 | 2570 | AAH03175.1 | |
D87953.1 | 16 | 2575 | BAA13505.1 | |
X92845.1 | 16 | 2563 | CAA63430.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |