Assay Details
Target Gene Details
Entrez Gene ID: | 375607 |
Gene Name: | N-acetyltransferase 16 (putative) |
Gene Aliases: |
C7orf52 |
Location: |
Chr.7:101170493-101180276 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NAT16 | NM_198571.2 | 2 | 403 | NP_940973.2 |
XM_005250325.1 | 3 | 382 | XP_005250382.1 | |
XM_011516168.1 | 2 | 364 | XP_011514470.1 | |
AK092119.1 | 2 | 364 | ||
AK096556.1 | 2 | 403 | BAC04815.1 | |
BC104841.1 | 3 | 382 | AAI04842.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2422376 | Chr.7:101100204 - 101373190 on Build GRCh38 | Duplication | MOGAT3 LOC101927746 MIR4653 AP1S1 PLOD3 ZNHIT1 SERPINE1 COL26A1 VGF FIS1 LOC101927721 NAT16 IFT22 CLDN15 |
nsv1030604 | Chr.7:101074451 - 101580185 on Build GRCh38 | Gain | LINC01007 MOGAT3 LOC101927746 MIR4653 AP1S1 PLOD3 TRIM56 ZNHIT1 SERPINE1 COL26A1 VGF FIS1 LOC101927721 NAT16 IFT22 CLDN15 |
dgv11534n54 | Chr.7:101160859 - 101238359 on Build GRCh38 | Loss | MOGAT3 VGF AP1S1 PLOD3 ZNHIT1 NAT16 CLDN15 |
esv2752134 | Chr.7:101156850 - 101498586 on Build GRCh38 | Gain | MOGAT3 LOC101927746 MIR4653 AP1S1 PLOD3 ZNHIT1 COL26A1 VGF FIS1 LOC101927721 NAT16 IFT22 CLDN15 |
nsv607973 | Chr.7:101139179 - 101230554 on Build GRCh38 | Loss | MOGAT3 VGF MIR4653 AP1S1 PLOD3 ZNHIT1 NAT16 SERPINE1 |
esv2734888 | Chr.7:99975623 - 101290285 on Build GRCh38 | Deletion | ZKSCAN1 PCOLCE MIR4653 GIGYF1 AP1S1 ZCWPW1 LOC102724094 ZNHIT1 UFSP1 SERPINE1 MCM7 STAG3 LOC105375429 MUC3A C7orf61 ZAN STAG3L5P CLDN15 ACHE AZGP1 SAP25 MOGAT3 PPP1R35 MUC12 MIR93 LAMTOR4 GAL3ST4 STAG3L5P-PVRIG2P-PILRB AP4M1 FIS1 GPC2 EPHB4 MIR106B AZGP1P1 PILRA MIR6840 PLOD3 TRIM56 C7orf43 ZSCAN21 TRIP6 MIR4658 TFR2 FBXO24 MUC17 PILRB TAF6 VGF MIR6875 MBLAC1 PVRIG ZASP COPS6 MEPCE ACTL6B LRCH4 POP7 MIR25 CNPY4 MOSPD3 PVRIG2P SLC12A9 AGFG2 GATS EPO PMS2P1 ZNF3 NYAP1 SPDYE3 TSC22D4 PCOLCE-AS1 NAT16 SRRT GNB2 |
nsv831075 | Chr.7:101049016 - 101273737 on Build GRCh38 | Loss | MOGAT3 VGF MIR4653 FIS1 AP1S1 PLOD3 TRIM56 ZNHIT1 NAT16 SERPINE1 MUC17 CLDN15 |
nsv607974 | Chr.7:101149529 - 101328323 on Build GRCh38 | Loss | MOGAT3 VGF LOC101927746 MIR4653 FIS1 AP1S1 PLOD3 LOC101927721 ZNHIT1 NAT16 IFT22 CLDN15 |
nsv1018841 | Chr.7:101117129 - 101225018 on Build GRCh38 | Gain | MOGAT3 VGF MIR4653 AP1S1 PLOD3 ZNHIT1 NAT16 SERPINE1 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |