Assay Details
Target Gene Details
Entrez Gene ID: | 64333 |
Gene Name: | Rho GTPase activating protein 9 |
Gene Aliases: |
10C, RGL1 |
Location: |
Chr.12:57472255-57488814 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv428283 | Chr.12:57435950 - 57608085 on Build GRCh38 | Gain | MIR6758 INHBC MBD6 DTX3 MIR616 DCTN2 KIF5A PIP4K2C GLI1 MARS DDIT3 ARHGAP9 INHBE |
nsv559034 | Chr.12:57466149 - 57618450 on Build GRCh38 | Loss | MIR6758 MBD6 ARHGEF25 DTX3 LOC101927583 MIR616 DCTN2 KIF5A PIP4K2C GLI1 MARS DDIT3 ARHGAP9 |
nsv469411 | Chr.12:57450266 - 57527405 on Build GRCh38 | Loss | MIR6758 INHBC MBD6 GLI1 MARS DDIT3 MIR616 ARHGAP9 INHBE |
nsv482992 | Chr.12:56206217 - 57706217 on Build GRCh38 | Gain+Loss | MIR6758 PRIM1 ARHGEF25 ANKRD52 RNF41 TAC3 DTX3 CS SPRYD4 SLC26A10 DCTN2 NABP2 B4GALNT1 STAT6 DDIT3 INHBE BAZ2A HBCBP RDH16 MIP OS9 LRP1 PIP4K2C STAC3 NEMP1 MBD6 NACA GPR182 CNPY2 LOC101927583 MIR616 HSD17B6 MYO1A LRP1-AS RBMS2 SDR9C7 KIF5A PAN2 MIR1228 COQ10A NAB2 GLS2 SNORD59B R3HDM2 IL23A SNORA105C NDUFA4L2 INHBC SHMT2 APOF SNORD59A ATP5B SLC39A5 NXPH4 PTGES3 GLI1 MARS TIMELESS ARHGAP9 ZBTB39 STAT2 |
nsv559035 | Chr.12:57467189 - 57476865 on Build GRCh38 | Loss | GLI1 ARHGAP9 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |