Assay Details
Target Gene Details
Entrez Gene ID: | 10109 |
Gene Name: | actin related protein 2/3 complex subunit 2 |
Gene Aliases: |
ARC34, PNAS-139, PRO2446, p34-Arc |
Location: |
Chr.2:218217094-218254348 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ARPC2 | NM_005731.3 | NP_005722.1 | ||
NM_152862.2 | NP_690601.1 | |||
XM_011510471.1 | XP_011508773.1 | |||
XM_017003113.1 | XP_016858602.1 | |||
XM_017003114.1 | XP_016858603.1 | |||
XM_017003115.1 | XP_016858604.1 | |||
AF006085.1 | AAB64190.1 | |||
AF279893.1 | AAK07550.1 | |||
AK092520.1 | ||||
AK093625.1 | ||||
AK290268.1 | ||||
BC000590.2 | AAH00590.1 | |||
BG282181.1 | ||||
BG765898.1 | ||||
BT006898.1 | AAP35544.1 | |||
U50523.1 | AAC50874.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1013137 | Chr.2:218208654 - 218248378 on Build GRCh38 | Gain | ARPC2 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |