Assay Details
Target Gene Details
Entrez Gene ID: | 2137 |
Gene Name: | exostosin like glycosyltransferase 3 |
Gene Aliases: |
BOTV, EXTL1L, EXTR1, REGR, RPR |
Location: |
Chr.8:28701473-28753690 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EXTL3 | NM_001440.3 | 3 | 622 | NP_001431.1 |
NR_073468.1 | ||||
NR_073469.1 | 3 | 622 | ||
XM_011544440.2 | 2 | 398 | XP_011542742.1 | |
AB007042.1 | 3 | 459 | BAA24080.1 | |
AB011091.1 | 3 | 481 | ||
AF001690.1 | 3 | 467 | AAC39598.1 | |
AF029231.1 | 1 | 128 | AAD01877.1 | |
AF083551.1 | 3 | 459 | AAD42041.1 | |
BC006363.2 | AAH06363.1 | |||
BI911162.1 | 3 | 490 | ||
DA222815.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |