Assay Details
Target Gene Details
Entrez Gene ID: | 6418 |
Gene Name: | SET nuclear proto-oncogene |
Gene Aliases: |
2PP2A, I2PP2A, IGAAD, IPP2A2, PHAPII, TAF-I, TAF-IBETA |
Location: |
Chr.9:128683432-128696396 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SET | NM_001122821.1 | NP_001116293.1 | ||
NM_001248000.1 | NP_001234929.1 | |||
NM_001248001.1 | NP_001234930.1 | |||
NM_003011.3 | NP_003002.2 | |||
XM_017015013.1 | XP_016870502.1 | |||
XM_017015014.1 | XP_016870503.1 | |||
XM_017015015.1 | XP_016870504.1 | |||
XM_017015016.1 | XP_016870505.1 | |||
AK223556.1 | BAD97276.1 | |||
AK300609.1 | ||||
AK300794.1 | ||||
AK311346.1 | ||||
AK315313.1 | ||||
AY349172.1 | AAQ79833.1 | |||
BC032749.1 | AAH32749.1 | |||
BU543146.1 | ||||
CR536543.1 | CAG38780.1 | |||
CR542050.1 | CAG46847.1 | |||
D45198.1 | BAA08139.1 | |||
EF534308.1 | ||||
JF729356.1 | ||||
M93651.1 | AAA60318.1 | |||
U51924.1 | AAC50460.1 | |||
X75091.1 | CAA52982.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv982347 | Chr.9:128692603 - 128696451 on Build GRCh38 | Duplication | SET |
nsv825097 | Chr.9:128648939 - 128702348 on Build GRCh38 | Loss | SET WDR34 |
nsv615400 | Chr.9:128688840 - 128693624 on Build GRCh38 | Gain | SET |
nsv482132 | Chr.9:128683655 - 128696396 on Build GRCh38 | Gain | SET |
dgv1237n67 | Chr.9:128662299 - 128700553 on Build GRCh38 | Gain | SET |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |