Assay Details
Target Gene Details
Entrez Gene ID: | 55089 |
Gene Name: | solute carrier family 38 member 4 |
Gene Aliases: |
ATA3, NAT3, PAAT, SNAT4 |
Location: |
Chr.12:46764761-46832422 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC38A4 | NM_001143824.1 | 10 | 974 | NP_001137296.1 |
NM_018018.4 | 11 | 1166 | NP_060488.2 | |
XM_005268997.2 | 10 | 975 | XP_005269054.1 | |
AB055003.1 | 9 | 798 | BAB84090.1 | |
AF193836.1 | 10 | 954 | AAK71508.1 | |
AF305814.1 | 11 | 1131 | AAK43528.1 | |
AK001053.1 | BAA91481.1 | |||
AK291168.1 | 10 | 975 | ||
BC069819.1 | 10 | 1027 | AAH69819.1 | |
BC101827.1 | 9 | 811 | AAI01828.1 | |
BC104913.1 | 9 | 810 | AAI04914.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv826361 | Chr.12:46765762 - 46807111 on Build GRCh38 | Loss | SLC38A4 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |