Assay Details
Target Gene Details
Entrez Gene ID: | 6691 |
Gene Name: | serine peptidase inhibitor, Kazal type 2 |
Gene Aliases: |
HUSI-II |
Location: |
Chr.4:56809860-56822210 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SPINK2 | NM_001271718.1 | NP_001258647.1 | ||
NM_001271719.1 | NP_001258648.1 | |||
NM_001271720.1 | NP_001258649.1 | |||
NM_001271721.1 | NP_001258650.1 | |||
NM_001271722.1 | NP_001258651.1 | |||
NM_021114.3 | NP_066937.1 | |||
NR_073417.1 | ||||
NR_073418.1 | ||||
NR_073419.1 | ||||
XM_011534405.1 | XP_011532707.1 | |||
AA776754.1 | ||||
AA973772.1 | ||||
AK312222.1 | ||||
AV743457.1 | ||||
AW592089.1 | ||||
BC022514.1 | AAH22514.1 | |||
BF589674.1 | ||||
BI520020.1 | ||||
BI561149.1 | ||||
BI562269.1 | ||||
BU853655.1 | ||||
CR542135.1 | CAG46932.1 | |||
DB451710.1 | ||||
GU727622.1 | ||||
HY020998.1 | ||||
HY028205.1 | ||||
HY043430.1 | ||||
HY055610.1 | ||||
X57655.1 | CAB37834.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv5214n100 | Chr.4:56786612 - 56999637 on Build GRCh38 | Gain | POLR2B NOA1 REST SPINK2 |
nsv1161088 | Chr.4:56678187 - 56838222 on Build GRCh38 | Duplication | HOPX SPINK2 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |