Assay Details
Target Gene Details
Entrez Gene ID: | 64689 |
Gene Name: | golgi reassembly stacking protein 1 |
Gene Aliases: |
GOLPH5, GRASP65, P65 |
Location: |
Chr.3:39096599-39108363 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GORASP1 | NM_001278789.1 | 7 | 2879 | NP_001265718.1 |
NM_001278790.1 | 6 | 2699 | NP_001265719.1 | |
NM_031899.3 | 9 | 3627 | NP_114105.1 | |
NR_103866.1 | 9 | 3123 | ||
NR_103867.1 | 9 | 3115 | ||
XM_006713301.3 | 9 | 3727 | XP_006713364.1 | |
XM_011534020.2 | 9 | 3637 | XP_011532322.1 | |
XM_011534021.2 | 8 | 3547 | XP_011532323.1 | |
XM_017007050.1 | 9 | 2813 | XP_016862539.1 | |
XM_017007051.1 | 8 | 3162 | XP_016862540.1 | |
XM_017007052.1 | 8 | 2758 | XP_016862541.1 | |
AI824080.1 | 1 | 163 | ||
AJ409349.1 | 9 | 2903 | CAC35160.1 | |
AK001574.1 | 8 | 3145 | ||
AK057043.1 | 6 | 2437 | ||
AK091168.1 | 10 | 2558 | BAC03598.1 | |
AK124755.1 | 9 | 3627 | ||
AW082955.1 | 1 | 155 | ||
BC008928.2 | 9 | 2844 | AAH08928.1 | |
BC075854.1 | 9 | 2913 | AAH75854.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs117482778] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
Intragenic Exonic non-DGV Variation |