Assay Details
Target Gene Details
Entrez Gene ID: | 9091 |
Gene Name: | phosphatidylinositol glycan anchor biosynthesis class Q |
Gene Aliases: |
GPI1, c407A10.1 |
Location: |
Chr.16:569968-584136 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PIGQ | NM_004204.3 | NP_004195.2 | ||
NM_148920.2 | NP_683721.1 | |||
AB003723.1 | BAA24948.1 | |||
AF030177.1 | AAC32661.1 | |||
AK300297.1 | ||||
AK314361.1 | ||||
BC006377.2 | AAH06377.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833113 | Chr.16:506513 - 628160 on Build GRCh38 | Loss | MIR5587 MIR3176 LOC107987233 LOC101929280 RAB11FIP3 CAPN15 PIGQ RAB40C NHLRC4 PRR35 LINC00235 |
esv3637579 | Chr.16:574322 - 585512 on Build GRCh38 | Loss | PIGQ |
esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication | GFER LOC105371045 LMF1 WDR90 LOC105371047 NHLRC4 STUB1 TPSAB1 TMEM204 MSRB1 MIR6859-4 LOC107984876 PKD1 NTN3 TBL3 UBE2I CLCN7 UNKL RHBDL1 RHOT2 PDPK1 RHBDF1 TRAF7 HS3ST6 SNORA78 LOC106660606 LINC00254 LOC105371184 LOC105371046 PRR35 RGS11 RPL23AP5 MIR1225 NTHL1 TELO2 SNHG19 LOC107987233 MIR6767 ECI1 ABCA3 RAB11FIP3 C1QTNF8 SNORA10 MIR4516 PTX4 DECR2 ARHGDIG MIR6511B1 WFIKKN1 MRPL28 KCTD5 MEIOB NME3 MPG RAB26 NDUFB10 MCRIP2 SNORA64 LOC105371049 FBXL16 HN1L PRR25 TBC1D24 ZNF598 MIR3177 IGFALS MLST8 IFT140 RAB40C LOC729652 NOXO1 SPSB3 RNF151 CASKIN1 HBA1 HBA2 WASIR2 HBM BAIAP3 HAGH LOC652276 MSLN RPS2 CAPN15 FLJ42627 FAM234A PIGQ SNHG9 ATP6V0C ERVK13-1 AMDHD2 MIR6768 LOC100134368 PRSS29P SOX8 WDR24 FAM173A RPL3L DNASE1L2 MIR3180-5 MIR4717 GNPTG JMJD8 RNPS1 NME4 SNORD60 LMF1-AS1 MIR3677 LOC105371038 EME2 FAHD1 MAPK8IP3 C16orf59 SNRNP25 C16orf91 CCNF LOC100287175 NPW MIR3176 PDIA2 CACNA1H TSC2 HBZ NARFL SSTR5 LINC00235 MIR5587 HAGHL PGP HBQ1 GNG13 MRPS34 TMEM8A TPSD1 E4F1 DDX11L10 CRAMP1 SLC9A3R2 PRSS27 MIR940 TPSB2 AXIN1 SYNGR3 NPRL3 POLR3K CCDC78 LOC101929280 MIR3178 SSTR5-AS1 NUBP2 CHTF18 RPUSD1 TPSG1 MIR662 TSR3 CEMP1 METTL26 BRICD5 ABCA17P LUC7L CCDC154 METRN |
nsv471063 | Chr.16:520623 - 852322 on Build GRCh38 | Loss | LOC100287175 MIR3176 LOC107987233 RAB11FIP3 WDR90 NARFL NHLRC4 STUB1 WFIKKN1 LINC00235 MIR5587 HAGHL MSLN GNG13 CAPN15 PIGQ MCRIP2 FBXL16 RHBDL1 PRR25 RHOT2 CCDC78 LOC101929280 WDR24 FAM173A CHTF18 RPUSD1 RAB40C LOC105371184 MIR662 PRR35 JMJD8 METTL26 LOC105371038 METRN |
nsv526903 | Chr.16:451362 - 810673 on Build GRCh38 | Loss | LOC100287175 MIR3176 LOC107987233 RAB11FIP3 WDR90 NARFL NHLRC4 STUB1 WFIKKN1 LINC00235 MIR5587 HAGHL MSLN GNG13 CAPN15 PIGQ MCRIP2 FBXL16 RHBDL1 PRR25 RHOT2 CCDC78 LOC101929280 WDR24 FAM173A CHTF18 RPUSD1 RAB40C LOC105371184 MIR662 PRR35 JMJD8 METTL26 LOC105371038 METRN |
nsv952898 | Chr.16:495001 - 1132900 on Build GRCh38 | Deletion | LOC100287175 MIR3176 LOC107987233 RAB11FIP3 LMF1 C1QTNF8 WDR90 NARFL SSTR5 NHLRC4 STUB1 WFIKKN1 LINC00235 MIR5587 HAGHL MSLN LOC107984876 GNG13 CAPN15 PIGQ MCRIP2 FBXL16 RHBDL1 PRR25 RHOT2 CCDC78 LOC101929280 SSTR5-AS1 SOX8 WDR24 FAM173A CHTF18 RPUSD1 RAB40C LOC105371184 MIR662 PRR35 JMJD8 METTL26 LMF1-AS1 LOC105371038 METRN |
dgv4823n54 | Chr.16:562698 - 662867 on Build GRCh38 | Loss | LOC100287175 METTL26 LOC107987233 LOC101929280 PIGQ LOC105371038 RAB40C WDR90 MCRIP2 NHLRC4 PRR35 WFIKKN1 |
nsv457304 | Chr.16:554554 - 611142 on Build GRCh38 | Gain | LOC107987233 LOC101929280 CAPN15 PIGQ RAB40C NHLRC4 PRR35 |
nsv1160325 | Chr.16:506925 - 836376 on Build GRCh38 | Duplication | LOC100287175 MIR3176 LOC107987233 RAB11FIP3 WDR90 NARFL NHLRC4 STUB1 WFIKKN1 LINC00235 MIR5587 HAGHL MSLN GNG13 CAPN15 PIGQ MCRIP2 FBXL16 RHBDL1 PRR25 RHOT2 CCDC78 LOC101929280 WDR24 FAM173A CHTF18 RPUSD1 RAB40C LOC105371184 MIR662 PRR35 JMJD8 METTL26 LOC105371038 METRN |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |