Assay Details
Target Gene Details
Entrez Gene ID: | 6503 |
Gene Name: | Src-like-adaptor |
Gene Aliases: |
SLA1, SLAP |
Location: |
Chr.8:133036728-133103360 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLA | NM_001045556.2 | 5 | 1034 | NP_001039021.1 |
NM_001045557.2 | 4 | 756 | NP_001039022.2 | |
NM_001282964.1 | 4 | 756 | NP_001269893.1 | |
NM_001282965.1 | 3 | 655 | NP_001269894.1 | |
NM_006748.3 | 3 | 400 | NP_006739.2 | |
XM_017013739.1 | 4 | 1227 | XP_016869228.1 | |
AJ238591.1 | CAB53536.1 | |||
AK297423.1 | 4 | 302 | ||
AK297519.1 | 3 | 191 | ||
AK309401.1 | 4 | 382 | ||
AK310092.1 | 4 | 339 | ||
AK310115.1 | 4 | 417 | ||
AK311010.1 | 4 | 343 | ||
AK312584.1 | 4 | 566 | ||
BC007042.1 | 3 | 341 | AAH07042.1 | |
BT019994.1 | 3 | 202 | AAV38797.1 | |
BX648382.1 | 5 | 617 | ||
CR536537.1 | 3 | 202 | CAG38774.1 | |
D89077.1 | 3 | 243 | BAA13758.1 | |
DA999385.1 | 3 | 400 | ||
DB265860.1 | 4 | 417 | ||
U30473.1 | 4 | 367 | AAC50357.1 | |
U44403.1 | 4 | 521 | AAC27662.1 |
Target Gene Details
Entrez Gene ID: | 7038 |
Gene Name: | thyroglobulin |
Gene Aliases: |
AITD3, TGN |
Location: |
Chr.8:132866943-133134902 on Build GRCh38 |
Assay Gene Location: | Within Intron 44 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TG | NM_003235.4 | NP_003226.4 | ||
XM_005251038.4 | XP_005251095.1 | |||
XM_006716622.3 | XP_006716685.1 | |||
XM_017013793.1 | XP_016869282.1 | |||
XM_017013794.1 | XP_016869283.1 | |||
XM_017013795.1 | XP_016869284.1 | |||
XM_017013796.1 | XP_016869285.1 | |||
XM_017013797.1 | XP_016869286.1 | |||
XM_017013798.1 | XP_016869287.1 | |||
AB209159.1 | BAD92396.1 | |||
AK299197.1 | ||||
AK303666.1 | ||||
AK303667.1 | ||||
BC127887.1 | ||||
BC140933.1 | ||||
U93033.2 | AAC51924.1 | |||
X05615.1 | CAA29104.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv482993 | Chr.8:132994656 - 133137835 on Build GRCh38 | Loss | SLA MIR7848 TG |
nsv6402 | Chr.8:133043596 - 133072827 on Build GRCh38 | Insertion | SLA MIR7848 TG |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic DGV Variation |