Assay Details
Target Gene Details
Entrez Gene ID: | 8732 |
Gene Name: | RNA guanylyltransferase and 5'-phosphatase |
Gene Aliases: |
CAP1A, HCE, HCE1, hCAP |
Location: |
Chr.6:88609897-88963716 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RNGTT | NM_001286426.1 | 15 | 1890 | NP_001273355.1 |
NM_001286428.1 | 14 | 1780 | NP_001273357.1 | |
NM_003800.4 | 16 | 1959 | NP_003791.3 | |
XM_017011401.1 | 17 | 2064 | XP_016866890.1 | |
AB009022.1 | BAA25894.1 | |||
AB009023.1 | BAA25895.1 | |||
AB012142.1 | 16 | 1894 | BAA25198.1 | |
AF025654.1 | 16 | 2027 | AAB91559.1 | |
AK295719.1 | 17 | 1837 | ||
AK299777.1 | 14 | 1770 | ||
AK312407.1 | ||||
BC019954.1 | 16 | 1926 | AAH19954.1 | |
BX537450.1 | 3 | 3147 | CAD97693.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv6087n100 | Chr.6:88552751 - 88940524 on Build GRCh38 | Gain | RNGTT |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |