Assay Details
Target Gene Details
Entrez Gene ID: | 51292 |
Gene Name: | guanosine monophosphate reductase 2 |
Gene Aliases: |
GMPR 2 |
Location: |
Chr.14:24232417-24239242 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GMPR2 | NM_001002000.2 | NP_001002000.1 | ||
NM_001002001.2 | NP_001002001.1 | |||
NM_001002002.2 | NP_001002002.1 | |||
NM_001283021.1 | NP_001269950.1 | |||
NM_001283022.1 | 1 | 383 | NP_001269951.1 | |
NM_001283023.1 | NP_001269952.1 | |||
NM_016576.4 | 1 | 383 | NP_057660.2 | |
NR_104265.1 | ||||
XM_005267740.3 | XP_005267797.1 | |||
XM_005267741.3 | XP_005267798.1 | |||
XM_005267742.3 | XP_005267799.1 | |||
XM_006720165.2 | XP_006720228.1 | |||
XM_017021356.1 | XP_016876845.1 | |||
XM_017021357.1 | XP_016876846.1 | |||
XM_017021358.1 | XP_016876847.1 | |||
XM_017021359.1 | XP_016876848.1 | |||
AB032903.1 | BAA93080.1 | |||
AF086919.1 | AAP97149.1 | |||
AF135159.1 | AAG09132.1 | |||
AF419346.1 | AAN32701.1 | |||
AK226166.1 | ||||
AV660418.1 | ||||
BC003053.1 | 1 | 332 | AAH03053.1 | |
BC008021.1 | 1 | 331 | AAH08021.2 | |
BC009832.2 | AAH09832.1 | |||
BC093039.1 | AAH93039.1 | |||
BE003291.1 | ||||
BI259066.1 | 1 | 345 | ||
BP347823.1 | ||||
BX161436.1 | CAD61908.1 | |||
BX161518.1 | CAD61951.1 | |||
BX247993.1 | CAD62327.1 | |||
BX375671.2 | 1 | 354 | ||
CR457156.1 | CAG33437.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |