Assay Details
Target Gene Details
Entrez Gene ID: | 22850 |
Gene Name: | ADNP homeobox 2 |
Gene Aliases: |
ZNF508 |
Location: |
Chr.18:80109031-80140346 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ADNP2 | NM_014913.3 | 1 | 228 | NP_055728.1 |
XM_011525883.2 | XP_011524185.1 | |||
AK292566.1 | ||||
BC071589.1 | 1 | 228 | AAH71589.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv3413n100 | Chr.18:80023442 - 80257174 on Build GRCh38 | Gain | RBFA TXNL4A PARD6G-AS1 RBFADN ADNP2 PARD6G |
nsv577958 | Chr.18:79925520 - 80130825 on Build GRCh38 | Gain | PQLC1 HSBP1L1 RBFA TXNL4A RBFADN ADNP2 |
nsv428359 | Chr.18:79277548 - 80255221 on Build GRCh38 | Gain | NFATC1 LOC107985149 HSBP1L1 RBFA LOC284240 KCNG2 TXNL4A PARD6G-AS1 ATP9B LOC284241 LOC105376875 CTDP1 PQLC1 RBFADN ADNP2 PARD6G |
nsv1065902 | Chr.18:78538045 - 80257174 on Build GRCh38 | Gain | NFATC1 LOC107985149 HSBP1L1 RBFA LOC284240 KCNG2 TXNL4A PARD6G-AS1 ATP9B LOC284241 LOC105376875 CTDP1 LOC100996274 PQLC1 SALL3 RBFADN ADNP2 PARD6G |
esv3643330 | Chr.18:80099479 - 80182196 on Build GRCh38 | Loss | PARD6G-AS1 ADNP2 PARD6G |
nsv1062512 | Chr.18:80081190 - 80127942 on Build GRCh38 | Gain | RBFADN ADNP2 |
nsv1061450 | Chr.18:79703531 - 80135801 on Build GRCh38 | Gain | PQLC1 HSBP1L1 RBFA KCNG2 TXNL4A RBFADN ADNP2 CTDP1 |
esv3643326 | Chr.18:79799861 - 80157032 on Build GRCh38 | Loss | PQLC1 HSBP1L1 RBFA KCNG2 TXNL4A PARD6G-AS1 RBFADN ADNP2 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |