Assay Details
Target Gene Details
Entrez Gene ID: | 3162 |
Gene Name: | heme oxygenase 1 |
Gene Aliases: |
HMOX1D, HO-1, HSP32, bK286B10 |
Location: |
Chr.22:35381067-35394214 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HMOX1 | NM_002133.2 | 3 | 509 | NP_002124.1 |
AF051791.1 | 2 | 381 | ||
AK313120.1 | 3 | 479 | ||
BC001491.2 | 3 | 468 | AAH01491.1 | |
BP288767.1 | 3 | 509 | ||
BT019785.1 | 2 | 402 | AAV38588.1 | |
CR456505.1 | 3 | 517 | CAG30391.1 | |
CR541945.1 | 3 | 402 | CAG46743.1 | |
CR541968.1 | 3 | 402 | CAG46766.1 | |
JF323038.1 | 3 | 406 | ||
X06985.1 | 3 | 482 | CAA30045.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |