Assay Details
Target Gene Details
Entrez Gene ID: | 79924 |
Gene Name: | adrenomedullin 2 |
Gene Aliases: |
AM2, dJ579N16.4 |
Location: |
Chr.22:50481724-50486440 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ADM2 | NM_001253845.1 | 3 | 1896 | NP_001240774.1 |
XM_017028940.1 | 2 | 1914 | XP_016884429.1 | |
AK090635.1 | 2 | 1914 | ||
AK092640.1 | 1 | 314 |
Target Gene Details
Entrez Gene ID: | 55586 |
Gene Name: | myo-inositol oxygenase |
Gene Aliases: |
ALDRL6 |
Location: |
Chr.22:50483988-50490321 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MIOX | XM_011530705.1 | 1 | 314 | XP_011529007.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv3669 | Chr.22:50475511 - 50509415 on Build GRCh38 | Insertion | ADM2 MIOX LMF2 NCAPH2 |
nsv1160782 | Chr.22:50463961 - 50571524 on Build GRCh38 | Deletion | TYMP SYCE3 SCO2 CPT1B CHKB-CPT1B ADM2 KLHDC7B MIOX ODF3B SBF1 LMF2 NCAPH2 |
nsv1057656 | Chr.22:50478783 - 50595520 on Build GRCh38 | Gain | TYMP SYCE3 SCO2 CPT1B CHKB-CPT1B ADM2 CHKB-AS1 KLHDC7B NCAPH2 CHKB MIOX ODF3B LMF2 |
nsv834238 | Chr.22:50402690 - 50556965 on Build GRCh38 | Loss | PPP6R2 TYMP SYCE3 SCO2 ADM2 LOC107985556 KLHDC7B MIOX ODF3B SBF1 LMF2 NCAPH2 |
nsv471224 | Chr.22:50435189 - 50580482 on Build GRCh38 | Loss | TYMP SYCE3 SCO2 CPT1B CHKB-CPT1B ADM2 KLHDC7B SBF1 NCAPH2 PPP6R2 CHKB MIOX ODF3B LMF2 |
nsv955201 | Chr.22:50421672 - 50537371 on Build GRCh38 | Deletion | PPP6R2 TYMP SCO2 ADM2 LOC107985556 MIOX ODF3B SBF1 LMF2 NCAPH2 |
nsv829338 | Chr.22:50440870 - 50489635 on Build GRCh38 | Gain | PPP6R2 ADM2 MIOX SBF1 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |