Assay Details
Target Gene Details
Entrez Gene ID: | 2050 |
Gene Name: | EPH receptor B4 |
Gene Aliases: |
HTK, MYK1, TYRO11 |
Location: |
Chr.7:100802565-100827521 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 16 - Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EPHB4 | NM_004444.4 | NP_004435.3 | ||
XM_017011816.1 | XP_016867305.1 | |||
AB209644.1 | BAD92881.1 | |||
AK301224.1 | ||||
AY056048.1 | AAL14195.1 | |||
BC004264.1 | AAH04264.1 | |||
BC052804.1 | AAH52804.1 | |||
U07695.1 | AAA20598.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2759547 | Chr.7:100757867 - 101105965 on Build GRCh38 | Gain+Loss | UFSP1 MUC3A MUC12 ACHE TRIM56 ZAN SLC12A9 MUC17 SRRT LOC102724094 EPHB4 TRIP6 MIR6875 |
nsv470381 | Chr.7:100642568 - 100834358 on Build GRCh38 | Loss | LOC105375429 ZAN GIGYF1 POP7 GNB2 EPO EPHB4 ACTL6B TFR2 |
esv2734888 | Chr.7:99975623 - 101290285 on Build GRCh38 | Deletion | PCOLCE STAG3L5P PMS2P1 UFSP1 TRIM56 MIR4658 NAT16 ZNHIT1 PLOD3 ZAN SPDYE3 GATS AZGP1 ZSCAN21 SAP25 TRIP6 ZNF3 MIR6840 MIR106B ZCWPW1 LOC105375429 AGFG2 POP7 PILRB MUC12 PPP1R35 ACHE MEPCE FIS1 MOSPD3 SERPINE1 STAG3L5P-PVRIG2P-PILRB ZKSCAN1 NYAP1 SRRT GNB2 C7orf61 AZGP1P1 MIR6875 MCM7 PVRIG2P COPS6 MOGAT3 GPC2 LAMTOR4 GIGYF1 SLC12A9 EPO LOC102724094 FBXO24 AP4M1 MIR25 PILRA MUC3A MIR4653 ZASP AP1S1 STAG3 GAL3ST4 TSC22D4 TAF6 MIR93 MBLAC1 PVRIG CNPY4 MUC17 CLDN15 LRCH4 EPHB4 ACTL6B VGF C7orf43 PCOLCE-AS1 TFR2 |
nsv607965 | Chr.7:100776465 - 100899511 on Build GRCh38 | Loss | ZAN UFSP1 SLC12A9 SRRT EPHB4 TRIP6 ACHE MIR6875 |
nsv1020837 | Chr.7:100715499 - 100911015 on Build GRCh38 | Loss | ZAN UFSP1 SLC12A9 SRRT EPO EPHB4 TRIP6 ACHE MIR6875 |
nsv951388 | Chr.7:100741378 - 100838178 on Build GRCh38 | Deletion | ZAN EPHB4 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |