Assay Details
Target Gene Details
Entrez Gene ID: | 55677 |
Gene Name: | IWS1, SUPT6H interacting protein |
Gene Aliases: |
- |
Location: |
Chr.2:127480807-127526886 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
IWS1 | NM_017969.2 | NP_060439.2 | ||
XM_005263707.3 | XP_005263764.1 | |||
XM_006712626.3 | XP_006712689.1 | |||
XM_011511450.1 | XP_011509752.1 | |||
XM_017004467.1 | XP_016859956.1 | |||
XM_017004468.1 | XP_016859957.1 | |||
AK001717.1 | BAA91858.1 | |||
AK027561.1 | BAB55198.1 | |||
AK056881.1 | BAB71301.1 | |||
AK294669.1 | ||||
AK296848.1 | ||||
AL834178.1 | CAD38875.1 | |||
BC017012.2 | AAH17012.3 | |||
BC110536.1 | AAI10537.1 | |||
BC110537.1 | AAI10538.1 |
More Information
Set Membership: |
Intragenic Non-exonic non-DGV Variation |