Assay Details
Target Gene Details
Entrez Gene ID: | 150223 |
Gene Name: | YdjC homolog (bacterial) |
Gene Aliases: |
- |
Location: |
Chr.22:21628089-21630326 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
YDJC | NM_001017964.1 | 1 | 87 | NP_001017964.1 |
XM_011529906.2 | 1 | 362 | XP_011528208.1 | |
BC110075.1 | 1 | 71 | AAI10076.1 | |
BC133043.1 | 1 | 182 | ||
BC133047.1 | 1 | 182 | ||
BI597711.1 | 1 | 106 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1160730 | Chr.22:21373849 - 21633730 on Build GRCh38 | Deletion | RIMBP3C RIMBP3B YDJC CCDC116 HIC2 PI4KAP2 UBE2L3 TMEM191C |
nsv516806 | Chr.22:21628603 - 21642781 on Build GRCh38 | Loss | YDJC CCDC116 SDF2L1 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |