Assay Details
Target Gene Details
Entrez Gene ID: | 7301 |
Gene Name: | TYRO3 protein tyrosine kinase |
Gene Aliases: |
BYK, Dtk, Etk-2, RSE, Rek, Sky, Tif |
Location: |
Chr.15:41559022-41579338 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TYRO3 | NM_006293.3 | NP_006284.2 | ||
XM_017022542.1 | XP_016878031.1 | |||
XM_017022543.1 | XP_016878032.1 | |||
AB209431.1 | BAD92668.1 | |||
AK295496.1 | ||||
BC029925.2 | AAH29925.2 | |||
BC049368.1 | AAH49368.1 | |||
BC051756.1 | AAH51756.1 | |||
D17517.1 | BAA04467.1 | |||
D50479.1 | BAA21781.1 | |||
U02566.1 | AAA17407.1 | |||
U05682.1 | AAA19236.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1138359 | Chr.15:41570340 - 41570603 on Build GRCh38 | Deletion | TYRO3 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |