Assay Details
Target Gene Details
Entrez Gene ID: | 6303 |
Gene Name: | spermidine/spermine N1-acetyltransferase 1 |
Gene Aliases: |
DC21, KFSD, KFSDX, SAT, SSAT, SSAT-1 |
Location: |
Chr.X:23783158-23786223 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SAT1 | NM_002970.3 | 1 | 140 | NP_002961.1 |
NR_027783.2 | 1 | 140 | ||
AF251292.1 | 1 | 150 | AAG44592.1 | |
AK310078.1 | 1 | 125 | ||
AK310094.1 | 1 | 121 | ||
AK312162.1 | 1 | 123 | ||
AK315888.1 | 1 | 63 | ||
AL050290.1 | 1 | 101 | ||
BC002503.2 | 1 | 101 | AAH02503.1 | |
BC008424.1 | 1 | 125 | AAH08424.1 | |
CD516700.1 | 1 | 146 | ||
CD638929.1 | 1 | 128 | ||
CN271682.1 | 1 | 140 | ||
M77693.1 | 1 | 111 | AAA60573.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv470346 | Chr.X:23738748 - 23786754 on Build GRCh38 | Gain | SAT1 ACOT9 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |