Assay Details
Target Gene Details
Entrez Gene ID: | 51192 |
Gene Name: | chemokine-like factor |
Gene Aliases: |
C32, CKLF1, CKLF2, CKLF3, CKLF4, HSPC224, UCK-1 |
Location: |
Chr.16:66552563-66566287 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CKLF | NM_001040138.2 | NP_001035228.1 | ||
NM_016326.3 | NP_057410.1 | |||
NM_016951.3 | NP_058647.1 | |||
NM_181640.2 | NP_857591.1 | |||
NM_181641.2 | NP_857592.1 | |||
AA234074.1 | ||||
AF057306.1 | AAF21255.1 | |||
AF096895.2 | AAF06722.1 | |||
AF135380.2 | AAF19599.1 | |||
AF135381.2 | AAF19600.1 | |||
AF145216.2 | AAF19350.1 | |||
AF151058.1 | AAF36144.1 | |||
AK290307.1 | ||||
AK301240.1 | ||||
AY358999.1 | AAQ89358.1 | |||
BC004380.1 | AAH04380.1 | |||
BC091478.1 | AAH91478.1 | |||
BU189908.1 | ||||
DA113473.1 | 2 | 320 | ||
DA804889.1 |
Target Gene Details
Entrez Gene ID: | 100529251 |
Gene Name: | CKLF-CMTM1 readthrough |
Gene Aliases: |
- |
Location: |
Chr.16:66552563-66579135 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CKLF-CMTM1 | NM_001202509.1 | NP_001189438.1 | ||
NM_001204098.1 | NP_001191027.1 | |||
NM_001204099.1 | NP_001191028.1 | |||
BM929895.1 | ||||
FY211334.1 | ||||
FY212695.1 |
More Information
Set Membership: |
Intragenic Non-exonic non-DGV Variation |