Assay Details
Target Gene Details
Entrez Gene ID: | 4793 |
Gene Name: | NFKB inhibitor beta |
Gene Aliases: |
IKBB, TRIP9 |
Location: |
Chr.19:38899503-38908894 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NFKBIB | NM_001243116.1 | 5 | 669 | NP_001230045.1 |
NM_002503.4 | 5 | 911 | NP_002494.2 | |
NR_040515.1 | 5 | 855 | ||
XM_006723226.2 | 5 | 847 | XP_006723289.2 | |
XM_006723227.3 | 5 | 810 | XP_006723290.3 | |
AB209784.1 | 5 | 877 | BAD93021.1 | |
AI684894.1 | 2 | 333 | ||
AK057862.1 | 5 | 806 | BAB71599.1 | |
AK290569.1 | 5 | 862 | ||
BC007197.1 | 3 | 511 | AAH07197.1 | |
BC015528.1 | 5 | 842 | AAH15528.1 | |
BI520169.1 | 5 | 669 | ||
BM449613.1 | ||||
BQ440907.1 | 5 | 643 | ||
BT006743.1 | 5 | 808 | AAP35389.1 | |
BU150265.1 | 2 | 134 | ||
BU634404.1 | 2 | 341 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |