Assay Details
Target Gene Details
Entrez Gene ID: | 26065 |
Gene Name: | LSM14A, mRNA processing body assembly factor |
Gene Aliases: |
C19orf13, FAM61A, RAP55, RAP55A |
Location: |
Chr.19:34172428-34229515 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LSM14A | NM_001114093.1 | NP_001107565.1 | ||
NM_015578.2 | NP_056393.2 | |||
XM_005258719.4 | XP_005258776.1 | |||
XM_005258720.4 | XP_005258777.1 | |||
XM_005258721.4 | XP_005258778.1 | |||
XM_011526708.2 | XP_011525010.1 | |||
XM_017026575.1 | XP_016882064.1 | |||
XM_017026576.1 | XP_016882065.1 | |||
XM_017026577.1 | XP_016882066.1 | |||
AB069974.1 | BAC99045.1 | |||
AB069975.1 | BAC99046.1 | |||
AK027643.1 | BAB55259.1 | |||
AK300208.1 | ||||
AL834398.1 | CAD39060.2 | |||
BC016842.1 | AAH16842.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv3515n100 | Chr.19:34122371 - 34196904 on Build GRCh38 | Gain | LSM14A |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |