Assay Details
Target Gene Details
Entrez Gene ID: | 169792 |
Gene Name: | GLIS family zinc finger 3 |
Gene Aliases: |
NDH, ZNF515 |
Location: |
Chr.9:3824127-4310694 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GLIS3 | NM_001042413.1 | 7 | 2752 | NP_001035878.1 |
NM_152629.3 | 6 | 1768 | NP_689842.3 | |
XM_005251386.4 | 6 | 2311 | XP_005251443.1 | |
XM_005251387.4 | 6 | 2589 | XP_005251444.1 | |
XM_005251388.4 | 5 | 2103 | XP_005251445.1 | |
XM_011517763.2 | 7 | 2431 | XP_011516065.1 | |
XM_011517764.2 | 7 | 2261 | XP_011516066.1 | |
XM_011517765.2 | 7 | 2798 | XP_011516067.1 | |
XM_011517766.2 | 6 | 2840 | XP_011516068.1 | |
XM_011517767.2 | 6 | 2234 | XP_011516069.1 | |
XM_017014361.1 | 6 | 2539 | XP_016869850.1 | |
AB209404.1 | 4 | 361 | BAD92641.1 | |
AK075059.1 | ||||
AK096318.1 | 4 | 395 | ||
BC033899.2 | 6 | 1768 | AAH33899.2 | |
DQ438878.1 | 4 | 696 | ABE66435.1 | |
DQ438879.1 | 4 | 579 | ABE66436.1 | |
DQ438880.1 | 4 | 579 | ABE66437.1 | |
KU178885.1 | 5 | 1574 | ||
KU178886.1 | 5 | 1574 | ||
KU178887.1 |
Target Gene Details
Entrez Gene ID: | 84850 |
Gene Name: | GLIS3 antisense RNA 1 |
Gene Aliases: |
C9orf70 |
Location: |
Chr.9:3898646-3901248 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GLIS3-AS1 | NR_026663.1 | 1 | 135 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |