Assay Details
Target Gene Details
Entrez Gene ID: | 10587 |
Gene Name: | thioredoxin reductase 2 |
Gene Aliases: |
SELZ, TR, TR-BETA, TR3, TRXR2 |
Location: |
Chr.22:19875518-19941992 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TXNRD2 | NM_006440.4 | 18 | 1896 | NP_006431.2 |
AB019694.1 | 18 | 1707 | BAA77601.2 | |
AB019695.1 | 18 | 1801 | BAA77602.2 | |
AB209724.1 | 18 | 1674 | BAD92961.1 | |
AF044212.1 | 18 | 1717 | AAD25167.1 | |
AF106697.1 | 18 | 1896 | AAD19597.1 | |
AF166126.1 | 13 | 1781 | AAF21431.1 | |
AF166127.1 | 18 | 1801 | AAF21432.1 | |
AF171054.1 | 18 | 1714 | AAD51324.1 | |
AF201385.1 | 17 | 2936 | AAG47635.1 | |
AK094698.1 | 5 | 2483 | ||
AK097708.1 | 17 | 1632 | ||
BC007489.2 | 17 | 1591 | AAH07489.3 | |
BC117354.1 | 18 | 1676 | ||
R46611.1 | 1 | 232 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834129 | Chr.22:19705617 - 19919068 on Build GRCh38 | Loss | SEPT5-GP1BB GNB1L SEPT5 TBX1 TXNRD2 GP1BB C22orf29 |
esv3893434 | Chr.22:19470889 - 20343372 on Build GRCh38 | Gain | GNB1L LOC440792 CCDC188 ARVCF COMT LINC00896 MIR4761 TBX1 TXNRD2 TRMT2A GP1BB LINC00895 MIR185 LOC101927859 SEPT5 DGCR6L MIR1286 CLDN5 CDC45 MIR3618 MIR6816 SEPT5-GP1BB UFD1L DGCR8 MIR1306 C22orf29 LOC284865 TANGO2 RTN4R ZDHHC8 RANBP1 |
nsv828939 | Chr.22:19696032 - 20324465 on Build GRCh38 | Loss | GNB1L LOC440792 CCDC188 ARVCF COMT LINC00896 MIR4761 TBX1 TXNRD2 TRMT2A GP1BB MIR185 SEPT5 DGCR6L MIR1286 MIR3618 MIR6816 SEPT5-GP1BB DGCR8 MIR1306 C22orf29 LOC284865 TANGO2 RTN4R ZDHHC8 RANBP1 |
nsv828938 | Chr.22:19691069 - 20324465 on Build GRCh38 | Gain | GNB1L LOC440792 CCDC188 ARVCF COMT LINC00896 MIR4761 TBX1 TXNRD2 TRMT2A GP1BB MIR185 SEPT5 DGCR6L MIR1286 MIR3618 MIR6816 SEPT5-GP1BB DGCR8 MIR1306 C22orf29 LOC284865 TANGO2 RTN4R ZDHHC8 RANBP1 |
nsv515664 | Chr.22:19873352 - 19892114 on Build GRCh38 | Loss | TXNRD2 |
nsv834130 | Chr.22:19837923 - 19940567 on Build GRCh38 | Loss | GNB1L TXNRD2 C22orf29 |
esv3568260 | Chr.22:19777375 - 19881883 on Build GRCh38 | Loss | GNB1L TBX1 TXNRD2 C22orf29 |
esv3647279 | Chr.22:19719848 - 19878980 on Build GRCh38 | Gain | SEPT5-GP1BB GNB1L SEPT5 TBX1 TXNRD2 GP1BB C22orf29 |
nsv953025 | Chr.22:19869078 - 19937977 on Build GRCh38 | Deletion | TXNRD2 |
dgv7983n54 | Chr.22:19762002 - 20321870 on Build GRCh38 | Gain | GNB1L LOC440792 CCDC188 ARVCF COMT DGCR6L MIR1286 LINC00896 MIR3618 MIR6816 MIR4761 DGCR8 TBX1 TXNRD2 MIR1306 TRMT2A MIR185 C22orf29 LOC284865 TANGO2 RTN4R ZDHHC8 RANBP1 |
esv3575418 | Chr.22:18161776 - 19963846 on Build GRCh38 | Gain | USP18 GGT3P GNB1L CLTCL1 LINC01311 DGCR11 COMT LOC100996401 DGCR2 LOC100996432 MIR4761 LOC642643 DGCR14 TBX1 TXNRD2 LOC100996415 GP1BB LINC00895 SEPT5 FAM230A CLDN5 RIMBP3 LOC102725072 DGCR10 CDC45 LOC729461 SEPT5-GP1BB TMEM191B UFD1L MRPL40 PRODH TSSK2 SLC25A1 HIRA DGCR5 DGCR6 C22orf39 DGCR9 C22orf29 GSC2 PI4KAP1 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |