Assay Details
Target Gene Details
Entrez Gene ID: | 9229 |
Gene Name: | DLG associated protein 1 |
Gene Aliases: |
DAP-1, DAP-1-ALPHA, DAP-1-BETA, DAP1, DLGAP1A, DLGAP1B, GKAP, SAPAP1 |
Location: |
Chr.18:3496032-4455307 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 19 - Exon 20 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DLGAP1 | NM_001003809.2 | NP_001003809.1 | ||
NM_001242762.1 | NP_001229691.1 | |||
NM_001242763.1 | NP_001229692.1 | |||
NM_001242764.1 | NP_001229693.1 | |||
NM_001242766.1 | NP_001229695.1 | |||
NM_001308390.1 | NP_001295319.1 | |||
NM_004746.3 | NP_004737.2 | |||
XM_005258171.2 | XP_005258228.1 | |||
XM_005258172.1 | XP_005258229.1 | |||
XM_005258173.3 | XP_005258230.2 | |||
XM_005258174.4 | XP_005258231.1 | |||
XM_006722367.3 | XP_006722430.1 | |||
XM_017026080.1 | XP_016881569.1 | |||
XM_017026081.1 | XP_016881570.1 | |||
XM_017026083.1 | XP_016881572.1 | |||
AB000276.1 | BAA23257.1 | |||
AB000277.1 | BAA23258.1 | |||
AK294717.1 | ||||
AK294747.1 | ||||
AK294777.1 | ||||
AK299880.1 | ||||
BC136453.1 | ||||
BC136454.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1059696 | Chr.18:3494893 - 3572306 on Build GRCh38 | Gain | DLGAP1 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |