Assay Details
Target Gene Details
Entrez Gene ID: | 55830 |
Gene Name: | glycosyltransferase 8 domain containing 1 |
Gene Aliases: |
AD-017, MSTP139 |
Location: |
Chr.3:52694484-52706083 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GLT8D1 | NM_001010983.2 | 1 | 188 | NP_001010983.1 |
NM_001278280.1 | 1 | 188 | NP_001265209.1 | |
NM_001278281.1 | 1 | 188 | NP_001265210.1 | |
NM_018446.3 | 1 | 188 | NP_060916.1 | |
XM_017006857.1 | 1 | 152 | XP_016862346.1 | |
XM_017006858.1 | 1 | 152 | XP_016862347.1 | |
XM_017006859.1 | 1 | 152 | XP_016862348.1 | |
BG831844.1 | 1 | 137 | ||
BI559798.1 | 1 | 193 |
Target Gene Details
Entrez Gene ID: | 28972 |
Gene Name: | signal peptidase complex subunit 1 |
Gene Aliases: |
HSPC033, SPC1, SPC12, YJR010C-A |
Location: |
Chr.3:52705841-52708182 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SPCS1 | NM_014041.3 | 1 | 56 | NP_054760.3 |
AX775785.1 | 2 | 570 | CAE11592.1 | |
BG724036.1 | 1 | 60 | ||
BU070128.1 | ||||
DB459917.1 | 1 | 55 |
More Information
Additional Information:
For this assay, SNP(s) [rs72960229] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
Intragenic Exonic non-DGV Variation |