Assay Details
Target Gene Details
Entrez Gene ID: | 83706 |
Gene Name: | fermitin family member 3 |
Gene Aliases: |
KIND3, MIG-2, MIG2B, UNC112C, URP2, URP2SF |
Location: |
Chr.11:64206665-64223891 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FERMT3 | NM_031471.5 | 14 | 1873 | NP_113659.3 |
NM_178443.2 | 14 | 1885 | NP_848537.1 | |
XM_011545294.2 | 14 | 1897 | XP_011543596.1 | |
XM_011545295.2 | 13 | 1730 | XP_011543597.1 | |
XM_017018398.1 | 14 | 1881 | XP_016873887.1 | |
XM_017018399.1 | 13 | 1733 | XP_016873888.1 | |
AK093719.1 | 14 | 1818 | BAC04220.1 | |
AY093951.1 | 14 | 1819 | AAM19736.1 | |
AY093952.1 | 14 | 1831 | AAM19737.1 | |
BC004347.1 | 14 | 1817 | AAH04347.2 | |
BC013366.2 | 14 | 1817 | AAH13366.1 | |
BC015584.2 | 14 | 1812 | AAH15584.1 | |
BG491422.1 | 1 | 55 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1159904 | Chr.11:64150129 - 64279169 on Build GRCh38 | Deletion | DNAJC4 PLCB3 FKBP2 VEGFB LOC105369340 GPR137 TRPT1 FERMT3 NUDT22 MACROD1 BAD PPP1R14B STIP1 |
nsv951010 | Chr.11:63942829 - 64352128 on Build GRCh38 | Deletion | MIR7155 FKBP2 TRPT1 OTUB1 NAA40 KCNK4-TEX40 KCNK4 PRDX5 BAD STIP1 TRMT112 DNAJC4 PLCB3 VEGFB LOC105369340 GPR137 FERMT3 CCDC88B NUDT22 MACROD1 ESRRA PPP1R14B TEX40 COX8A FLRT1 |
dgv1963n54 | Chr.11:64174262 - 64329761 on Build GRCh38 | Loss | TRMT112 DNAJC4 PLCB3 FKBP2 VEGFB LOC105369340 GPR137 TRPT1 FERMT3 KCNK4-TEX40 KCNK4 PRDX5 NUDT22 ESRRA BAD PPP1R14B TEX40 STIP1 |
nsv555187 | Chr.11:64185649 - 64260416 on Build GRCh38 | Loss | DNAJC4 PLCB3 FKBP2 NUDT22 VEGFB LOC105369340 PPP1R14B TRPT1 STIP1 FERMT3 |
nsv832186 | Chr.11:64204788 - 64402587 on Build GRCh38 | Loss | MIR7155 TRMT112 DNAJC4 PLCB3 FKBP2 VEGFB LOC105369340 GPR137 TRPT1 FERMT3 KCNK4-TEX40 KCNK4 CCDC88B PRDX5 RPS6KA4 NUDT22 ESRRA BAD PPP1R14B TEX40 MIR1237 |
nsv555184 | Chr.11:64156974 - 64275525 on Build GRCh38 | Loss | DNAJC4 PLCB3 FKBP2 VEGFB LOC105369340 GPR137 TRPT1 FERMT3 NUDT22 MACROD1 BAD PPP1R14B STIP1 |
nsv469961 | Chr.11:64055468 - 64318495 on Build GRCh38 | Loss | TRMT112 DNAJC4 PLCB3 FKBP2 VEGFB LOC105369340 GPR137 TRPT1 FERMT3 KCNK4-TEX40 KCNK4 PRDX5 NUDT22 MACROD1 ESRRA BAD PPP1R14B TEX40 STIP1 FLRT1 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |