Assay Details
Target Gene Details
Entrez Gene ID: | 716 |
Gene Name: | complement component 1, s subcomponent |
Gene Aliases: |
- |
Location: |
Chr.12:7060676-7071032 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
C1S | NM_001734.3 | 4 | 586 | NP_001725.1 |
NM_201442.2 | 4 | 673 | NP_958850.1 | |
XM_005253760.1 | 4 | 565 | XP_005253817.1 | |
AK025309.1 | 1 | 62 | ||
AK290295.1 | 4 | 620 | ||
AM886411.1 | 3 | 308 | ||
BC056903.1 | 4 | 613 | AAH56903.1 | |
CD358820.1 | 4 | 545 | ||
DB257339.1 | ||||
J04080.1 | 4 | 512 | AAA51852.1 | |
M18767.1 | 4 | 532 | AAA51853.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |