Assay Details
Target Gene Details
Entrez Gene ID: | 390212 |
Gene Name: | G protein-coupled receptor 152 |
Gene Aliases: |
PGR5 |
Location: |
Chr.11:67451301-67452729 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GPR152 | NM_206997.1 | 1 | 822 | NP_996880.1 |
AY569571.1 | 1 | 822 | AAS76893.1 | |
BC119779.1 | 1 | 822 | ||
BC122869.1 | 1 | 822 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv468604 | Chr.11:67418529 - 67516420 on Build GRCh38 | Loss | PITPNM1 PTPRCAP GPR152 CABP4 CARNS1 CDK2AP2 CORO1B AIP RPS6KB2 TMEM134 MIR6752 |
dgv1977n54 | Chr.11:67397024 - 67521123 on Build GRCh38 | Loss | PTPRCAP CDK2AP2 CORO1B TBC1D10C RPS6KB2 RAD9A TMEM134 PPP1CA MIR6752 PITPNM1 GPR152 CABP4 CARNS1 AIP CABP2 |
dgv52n111 | Chr.11:67362651 - 67521123 on Build GRCh38 | Deletion | PTPRCAP CDK2AP2 CORO1B TBC1D10C RPS6KB2 RAD9A TMEM134 PPP1CA MIR6752 PITPNM1 GPR152 CABP4 CARNS1 CLCF1 AIP LOC100130987 CABP2 |
nsv521723 | Chr.11:67408221 - 67543787 on Build GRCh38 | Loss | PTPRCAP CDK2AP2 CORO1B TBC1D10C RPS6KB2 TMEM134 MIR6752 PITPNM1 GPR152 CABP4 CARNS1 AIP CABP2 |
nsv527056 | Chr.11:67439955 - 67526837 on Build GRCh38 | Loss | PITPNM1 GPR152 CABP4 CDK2AP2 CORO1B AIP CABP2 TMEM134 MIR6752 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |